CAS RN 1401223-22-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
PI-1840, MedChemExpress
MedChemExpress PI-1840 is a potent and selective inhibitor for chymotrypsin-like (CT-L) (IC50 value = 27 (plusmn) 0.14 nM) over trypsin-like and peptidylglutamyl peptide hydrolyzing (IC50 values >100 (mu)M) activities of the proteasome.