CAS RN 139446-82-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
6'-O-Apiosyl-5-O-Methylvisammioside, MedChemExpress
MedChemExpress 6'-O-Apiosyl-5-O-Methylvisammioside is one of the components of the traditional Chinese medicine Kang-Jing.