CAS RN 138794-73-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Sardomozide dihydrochloride, MedChemExpress
MedChemExpress Sardomozide dihydrochloride is an S-adenosylmethionine decarboxylase (SAMDC) inhibitor with an IC50 of 5 nM.