CAS RN 13860-66-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
N3PT, MedChemExpress
MedChemExpress N3PT(N3-pyridyl thiamine) is a potent and selective transketolase(TKT) inhibitor (IC50= 22 nM for Apo-TK) both in vitro and in vivo.