CAS RN 1383716-33-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
Vps34-IN-1, MedChemExpress
MedChemExpress Vps34-IN-1 is an inhibitor of Vps34 extracted from patent WO2012085815A1, compound example 16a, with an IC50 of 4 nM. Vps34-IN-1 modulates autophagy.
2