CAS RN 1375603-38-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Seviteronel R enantiomer, MedChemExpress
MedChemExpress Seviteronel R enantiomer (VT-464 R enantiomer) is the R enantiomer of Seviteronel (VT-464), which is a potent CYP17 lyase inhibitor(h-Lyase IC50=69 nM); Seviteronel (VT-464) R enantiomer's activity is unknown.