CAS RN 1375603-36-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Seviteronel racemate, MedChemExpress
MedChemExpress Seviteronel racemate (VT-464 racemate) is the racemate form of Seviteronel (VT-464), which is a potent CYP17 lyase inhibitor(h-Lyase IC50=nM)inhibition.