CAS RN 1375105-96-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
FASN-IN-4, MedChemExpress
MedChemExpress FASN-IN-4 is a potent inhibitor of fatty acid synthase (FASN) with an IC50 of 10 nM (WO2012064642A1, compound 29). FASN-IN-4 also inhibits SARS-CoV-2 with an EC50 of 18.6 nM.