CAS RN 1373422-53-7
IUPAC Nombre:
3-{[2-(pyridin-2-yl)-6-(2,3,4,5-tetrahydro-1H-3-benzazepin-3-yl)pyrimidin-4-yl]amino}propanoic acid
Sinónimos:
3-{[2-(pyridin-2-yl)-6-(1,2,4,5-tetrahydro-3-benzazepin-3-yl)pyrimidin-4-yl]amino}propanoic acid
Peso molecular (g/mol):
389.46
Fórmula molecular:
C22H23N5O2
InChi Key:
AVZCPICCWKMZDT-UHFFFAOYSA-N
SMILES:
OC(=O)CCNC1=NC(=NC(=C1)N1CCC2=CC=CC=C2CC1)C1=CC=CC=N1
1
–
2
de
2
resultados
1
Tocris Bioscience™ GSK J1
CAS: 1373422-53-7 Fórmula molecular: C22H23N5O2
2
GSK-J1, MedChemExpress
MedChemExpress GSK-J1 is a potent inhibitor of H3K27me3/me2-demethylases JMJD3/KDM6B and UTX/KDM6A, with IC50 of 60 nM towards KDM6B.