CAS RN 137061-48-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
2
de
2
resultados
1
MedChemExpress PACAP (1-38), human, ovine, rat
MedChemExpress PACAP (1-38), human, ovine, rat is a neuropeptide with 38 amino acid residues. PACAP (1-38) binds to PACAP type I receptor, PACAP type II receptor VIP1, and PACAP type II receptor VIP2 with IC50s of 4 nM, 2 nM, and 1 nM, respectively.
2
MedChemExpress PACAP (1-38), human, ovine, rat
MedChemExpress PACAP (1-38), human, ovine, rat is a neuropeptide with 38 amino acid residues. PACAP (1-38) binds to PACAP type I receptor, PACAP type II receptor VIP1, and PACAP type II receptor VIP2 with IC50s of 4 nM, 2 nM, and 1 nM, respectively.