CAS RN 1370261-97-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
PRT062607 Hydrochloride, MedChemExpress
MedChemExpress PRT062607 Hydrochloride (P505-15 Hydrochloride) is a highly specific and potent inhibitor of purified Syk (IC50 1-2 nM).