CAS RN 1370003-76-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
YK11, MedChemExpress
MedChemExpress YK11 is a partial agonist of androgen receptor, with osteogenic activity.