CAS RN 136831-48-6
IUPAC Nombre:
(2Z)-3-(3,4-dimethoxyphenyl)-2-(pyridin-3-yl)prop-2-enenitrile
Sinónimos:
(2Z)-3-(3,4-dimethoxyphenyl)-2-(pyridin-3-yl)prop-2-enenitrile
Peso molecular (g/mol):
266.3
Fórmula molecular:
C16H14N2O2
InChi Key:
DBGZNJVTHYFQJI-RIYZIHGNSA-N
SMILES:
COC1=CC=C(\C=C(/C#N)C2=CC=CN=C2)C=C1OC
1
–
1
de
1
resultados
1
RG13022, MedChemExpress
MedChemExpress RG13022 is a tyrosine kinase inhibitor; inhibits the autophosphorylation reaction of the EGF receptor with an IC50 of 4 μM.