CAS RN 1361951-15-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
TP-3654, MedChemExpress
MedChemExpress TP-3654 is a second-generation Pim kinase inhibitor with Ki values of 5 and 42 nM for Pim-1 and Pim-3, respectively.