CAS RN 1361504-77-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
DG172 dihydrochloride, MedChemExpress
MedChemExpress DG172 dihydrochloride is a selective PPARβ/δ antagonist, with an IC50 of 27 nM.