CAS RN 1360614-48-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Necrosulfonamide, MedChemExpress
MedChemExpress Necrosulfonamide is a necroptosis inhibitor acting by selectively targeting the mixed lineage kinase domain-like protein (MLKL). Necrosulfonamide prevents MLKL-RIP1-RIP3 necrosome complex from interacting with its downstream effectors. MLKL is a critical substrate of RIP3 during the induction of necrosis.