CAS RN 1356962-34-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
HG-14-10-04, MedChemExpress
MedChemExpress HG-14-10-04 (example 10) is a potent and specific ALK inhibitor with an IC50 of 20 nM.