CAS RN 1354690-24-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Leniolisib, MedChemExpress
MedChemExpress Leniolisib (CDZ173) is a potent and selective PI3Kδ inhibitor. Leniolisib has the potential for immunodeficiency disorders treatment.