CAS RN 1354037-26-5
IUPAC Nombre:
ethyl 3-(2-amino-1-cyanoeth-1-en-1-yl)-6,7-dichloro-1-methyl-1H-indole-2-carboxylate
Sinónimos:
ethyl 3-(2-amino-1-cyanoeth-1-en-1-yl)-6,7-dichloro-1-methylindole-2-carboxylate
Peso molecular (g/mol):
338.19
Fórmula molecular:
C15H13Cl2N3O2
InChi Key:
CXJCGSPAPOTTSF-UHFFFAOYSA-N
SMILES:
CCOC(=O)C1=C(C(=CN)C#N)C2=CC=C(Cl)C(Cl)=C2N1C
1
–
1
de
1
resultados
1
KH-CB19, MedChemExpress
MedChemExpress KH-CB19 is a potent and highly specific inhibitor of the CDC2-like kinase isoforms 1 and 4 (CLK1/CLK4).