CAS RN 1353625-73-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Presatovir, MedChemExpress
MedChemExpress Presatovir (GS-5806) is an orally bioavailable RSV fusion inhibitor with a mean EC50 value of 0.43 nM.