CAS RN 135330-08-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
RP-54745, MedChemExpress
MedChemExpress RP-54745 is an inhibitor of macrophage stimulation and interleukin-1 production, and a potential antirheumatic compound.