CAS RN 1351635-67-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
ONO-4059 analog, MedChemExpress
MedChemExpress ONO-4059 analog is the analog of ONO-4059, ONO-4059 is a highly potent and selective Btk inhibitor.