CAS RN 13512-31-7
IUPAC Nombre:
methyl 2-{[(benzyloxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoate
Sinónimos:
methyl 2-{[(benzyloxy)carbonyl]amino}-3-(4-hydroxyphenyl)propanoate
Peso molecular (g/mol):
329.35
Fórmula molecular:
C18H19NO5
InChi Key:
SLLMDHBKALJDBW-UHFFFAOYNA-N
SMILES:
COC(=O)C(CC1=CC=C(O)C=C1)NC(=O)OCC1=CC=CC=C1
1
–
1
de
1
resultados
1
Éster metílico de N-benciloxicarbonil-L-tirosina, 98 %, Thermo Scientific Chemicals
CAS: 13512-31-7 Fórmula molecular: C18H19NO5 Peso molecular (g/mol): 329.35 Número MDL: MFCD00038784 Clave InChI: SLLMDHBKALJDBW-XISACWJONA-N Sinónimo: z-tyr-ome,z-l-tyrosine methyl ester,n-cbz-l-tyrosine methyl ester,s-methyl 2-benzyloxy carbonyl amino-3-4-hydroxyphenyl propanoate,cbz-tyr-ome,z-l-tyr-ome,z-tyr-och3,cbz-tyrosine methyl ester,n-cbz-l-tyrosinemethylester,n-benzyloxycarbonyltyrosine methyl ester PubChem CID: 7010613 SMILES: COC(=O)[C@H](CC1=CC=C(O)C=C1)NC(=O)OCC1=CC=CC=C1