CAS RN 1349723-93-8
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Elacestrant dihydrochloride, MedChemExpress
MedChemExpress Elacestrant dihydrochloride (RAD1901 dihydrochloride) is an orally available selective estrogen receptor degrader (SERD) with IC50s of 48 and 870 nM for ERα and ERβ, respectively.