CAS RN 1346753-00-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
YHO-13351, MedChemExpress
MedChemExpress YHO-13351 is the prodrug of YHO-13177, which is a potent and specific inhibitor of BCRP.