CAS RN 1346572-63-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
GSK503, MedChemExpress
MedChemExpress GSK503 is a potent and specific inhibitor of EZH2 methyltransferase with Kiapp values of 3 to 27 nM.