CAS RN 1345808-25-4
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Arginase inhibitor 1, MedChemExpress
MedChemExpress Arginase inhibitor 1 is a potent inhibitor of human arginases I and II with IC50s of 223 and 509 nM, respectively.