CAS RN 1345675-02-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
ETP-46464, MedChemExpress
MedChemExpress ETP-46464 is an effective mTOR and ATR inhibitor with IC50s of 0.6 and 14 nM, respectively.