CAS RN 134418-28-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Dehydroandrographolide, MedChemExpress
MedChemExpress Dehydroandrographolide is extracted from herbal medicine Andrographis paniculata Nees. Dehydroandrographolide reduces oxidative stress in LPS-induced acute lung injury by inactivating iNOS. Dehydroandrographolide has anti-infective activity.