CAS RN 133718-30-6
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
R 80123, MedChemExpress
MedChemExpress R 80123 is the Z-isomer of R 79595, is also a highly selective phosphodiesterase inhibitor.