CAS RN 133550-49-9
IUPAC Nombre:
2-(2,3-dihydro-1H-indole-1-carbonyl)-3-(3,4-dihydroxyphenyl)prop-2-enenitrile
Sinónimos:
2-(2,3-dihydroindole-1-carbonyl)-3-(3,4-dihydroxyphenyl)prop-2-enenitrile
Peso molecular (g/mol):
306.32
Fórmula molecular:
C18H14N2O3
InChi Key:
MYKCTDWWIWGLHW-UHFFFAOYSA-N
SMILES:
OC1=C(O)C=C(C=C(C#N)C(=O)N2CCC3=CC=CC=C23)C=C1
1
–
1
de
1
resultados
1
Tyrphostin AG 528, MedChemExpress
MedChemExpress Tyrphostin AG 528 is an inhibitor of EGFR and ErbB2 with IC50s of 4.9 and 2.1 μM, respectively. Tyrphostin AG 528 (Tyrphostin B66) is a protein tyrosine kinase inhibitor, with IC50s of 4.9 μM for epidermal growth factor receptors (EGFR) and 2.1 μM for ErbB2. Tyrphostin AG 528 is also an anticancer agent.