CAS RN 1334850-99-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Mito-TEMPO, MedChemExpress
MedChemExpress Mito-TEMPO is a mitochondria-targeted superoxide dismutase mimetic with superoxide and alkyl radical scavenging properties.