CAS RN 132996-61-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
3
de
3
resultados
1
MedChemExpress Osteogenic Growth Peptide, OGP
MedChemExpress Osteogenic Growth Peptide, OGP is a short, naturally occurring 14-mer growth factor peptide found in serum at μM concentrations.
2
MedChemExpress Osteogenic Growth Peptide, OGP
MedChemExpress Osteogenic Growth Peptide, OGP is a short, naturally occurring 14-mer growth factor peptide found in serum at μM concentrations.
3
MedChemExpress Osteogenic Growth Peptide, OGP
MedChemExpress Osteogenic Growth Peptide, OGP is a short, naturally occurring 14-mer growth factor peptide found in serum at μM concentrations.