CAS RN 1315329-43-1
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
HG6-64-1, MedChemExpress
MedChemExpress HG6-64-1 is a potent and selective B-Raf inhibitor extracted from patent WO 2011090738 A2, example 9 (XI-1); has a IC50 of 0.09 μM on B-raf V600E transformed Ba/F3 cells.