CAS RN 131436-22-1
IUPAC Nombre:
(1S)-12-acetyl-3,5,11-trihydroxy-1-methyl-13-oxo-8-oxatricyclo[7.4.0.0²,⁷]trideca-2,4,6,9,11-pentaene-6-carboxamide
Sinónimos:
(1S)-12-acetyl-3,5,11-trihydroxy-1-methyl-13-oxo-8-oxatricyclo[7.4.0.0²,⁷]trideca-2,4,6,9,11-pentaene-6-carboxamide
Peso molecular (g/mol):
331.28
Fórmula molecular:
C16H13NO7
InChi Key:
GEWLYFZWVLXQME-MRXNPFEDSA-N
SMILES:
CC(=O)C1=C(O)C=C2OC3=C(C(N)=O)C(O)=CC(O)=C3[C@]2(C)C1=O
1
–
2
de
2
resultados
1
Cercosporamide, MedChemExpress
MedChemExpress Cercosporamide is a highly potent, ATP-competitive Pkc1 kinase inhibitor, with an IC50 of Ki of Mnk inhibitor.
2
Tocris Bioscience™ Cercosporamide
Potent Mnk2 inhibitor