CAS RN 1313363-54-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
NVP-CGM097, MedChemExpress
MedChemExpress NVP-CGM097 is a potent and selective MDM2 inhibitor with IC50 of 1.7±0.1 nM for hMDM2.