CAS RN 1310340-58-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
VBY-825, MedChemExpress
MedChemExpress VBY-825 is a novel, reversible cathepsin inhibitor with high potency against cathepsins B, L, S and V.