CAS RN 130493-03-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Bimoclomol, MedChemExpress
MedChemExpress Bimoclomol is a heat shock protein (HSP) coinducer, used for treatment of cardiovascular diseases.