CAS RN 1303470-48-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
KHK-IN-1 hydrochloride, MedChemExpress
MedChemExpress KHK-IN-1 hydrochloride is a potent ketohexokinase (KHK) inhibitor with IC50 of 12 nM, interacts with Asp-27B in the ATP-binding region of KHK.