CAS RN 1300031-70-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Bromodomain inhibitor-8, MedChemExpress
MedChemExpress Bromodomain inhibitor-8 (Intermediate 21) is a BET bromodomain inhibitor for treating autoimmune and inflammatory diseases.