CAS RN 1286279-29-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
BMS-813160, MedChemExpress
MedChemExpress BMS-813160 is the first dual CCR2/CCR5 antagonist, has the potential for cardiovascular treatment.