CAS RN 1271738-62-5
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
Menin-MLL inhibitor MI-2, MedChemExpress
MedChemExpress Menin-MLL inhibitor MI-2 is a Menin-MLL interaction inhibitor with IC50 of 446±28 nM.