CAS RN 127-56-0
IUPAC Nombre:
sodium acetyl(4-aminobenzenesulfonyl)azanide
Sinónimos:
sulfacetamide sodium sodium sulfanilylacetamide sulfacetamide sodium anhydrous sodium N-sulfanilylacetamide
Peso molecular (g/mol):
236.22
Fórmula molecular:
C8H9N2NaO3S
InChi Key:
PQMSFAORUFMASU-UHFFFAOYSA-M
SMILES:
[Na+].CC(=O)[N-]S(=O)(=O)C1=CC=C(N)C=C1
1
–
1
de
1
resultados
1
Sulfacetamide Sodium, MedChemExpress
MedChemExpress Sulfacetamide Sodium is an anti-infective agent that is used topically to treat skin infections and orally for urinary tract infections.