CAS RN 1269815-17-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
CDK-IN-2, MedChemExpress
MedChemExpress CDK-IN-2 is a potent and specific CDK9 inhibitor with IC50 of <8 nM, extracted from reference 1, example 4.