CAS RN 1264191-73-2
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
VTP-27999 Hydrochloride, MedChemExpress
MedChemExpress VTP-27999 Hcl is an alkyl amine Renin inhibitor; VTP-27999 is useful for Hypertension and End-Organ Diseases.