CAS RN 1260530-25-3
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
GSK 525768A, MedChemExpress
MedChemExpress GSK 525768A is the inactive enantiomer of GSK525762A. GSK 525768A has no activity towards BET.