CAS RN 126-17-0
IUPAC Nombre:
(1S,2S,4S,5'R,6R,7S,8R,9S,12S,13R,16S)-5',7,9,13-tetramethyl-5-oxaspiro[pentacyclo[10.8.0.0²,⁹.0⁴,⁸.0¹³,¹⁸]icosane-6,2'-piperidin]-18-en-16-ol
Sinónimos:
solasodine (3β,22β,25s)-isomer solasodine purapuridine (3β,22β,25s)-solasodine solasod-5-en-3-β-ol solasod-5-en-3β-ol (8CI) (3β,22β,25S)-isomer solasodine solasod-5-en-3β-ol tomatidenol salasdine
Peso molecular (g/mol):
413.65
Fórmula molecular:
C27H43NO2
InChi Key:
KWVISVAMQJWJSZ-VKROHFNGSA-N
SMILES:
C[C@H]1[C@H]2[C@H](C[C@H]3[C@@H]4CC=C5C[C@@H](O)CC[C@]5(C)[C@H]4CC[C@]23C)O[C@]11CC[C@@H](C)CN1
1
–
1
de
1
resultados
1
Solasodine, MedChemExpress
MedChemExpress Solasodine (Purapuridine) is a steroidal alkaloid that occurs in plants of the Solanaceae family. Solasodine has neuroprotective, antifungal, hypotensive, anticancer, antiatherosclerotic, antiandrogenic and anti-inflammatory activities.