CAS RN 1256094-72-0
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
AI-10-49, MedChemExpress
MedChemExpress AI-10-49 is a protein-protein interaction inhibitor that selectively binds to CBFβ-SMMHC and disrupts its binding to RUNX1 with a FRET IC50 of 0.26 uM.