CAS RN 1255580-76-7
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
UNC0638, MedChemExpress
MedChemExpress UNC0638 selectively inhibits G9a and GLP histone methyltransferase activity with IC50s of less than 15 nM and 19 nM, respectively. UNC0638 has anti-FMDV (foot-and-mouth disease virus) and anti-VSV (vesicular stomatitis virus) activities.