CAS RN 1252679-52-9
IUPAC Nombre:
—
Sinónimos:
—
Peso molecular (g/mol):
—
Fórmula molecular:
—
InChi Key:
—
SMILES:
—
1
–
1
de
1
resultados
1
ITX5061, MedChemExpress
MedChemExpress ITX5061 is a type II inhibitor of p38 MAPK and also an antagonist of scavenger receptor B1 (SR-B1).